Merck
  • Identification of a sudden cardiac death susceptibility locus at 2q24.2 through genome-wide association in European ancestry individuals.
Identification of a sudden cardiac death susceptibility locus at 2q24.2 through genome-wide association in European ancestry individuals.
PLoS genetics (2011-07-09)
Dan E Arking, M Juhani Junttila, Philippe Goyette, Adriana Huertas-Vazquez, Mark Eijgelsheim, Marieke T Blom, Christopher Newton-Cheh, Kyndaron Reinier, Carmen Teodorescu, Audrey Uy-Evanado, Naima Carter-Monroe, Kari S Kaikkonen, Marja-Leena Kortelainen, Gabrielle Boucher, Caroline Lagacé, Anna Moes, XiaoQing Zhao, Frank Kolodgie, Fernando Rivadeneira, Albert Hofman, Jacqueline C M Witteman, André G Uitterlinden, Roos F Marsman, Raha Pazoki, Abdennasser Bardai, Rudolph W Koster, Abbas Dehghan, Shih-Jen Hwang, Pallav Bhatnagar, Wendy Post, Gina Hilton, Ronald J Prineas, Man Li, Anna Köttgen, Georg Ehret, Eric Boerwinkle, Josef Coresh, W H Linda Kao, Bruce M Psaty, Gordon F Tomaselli, Nona Sotoodehnia, David S Siscovick, Greg L Burke, Eduardo Marbán, Peter M Spooner, L Adrienne Cupples, Jonathan Jui, Karen Gunson, Y Antero Kesäniemi, Arthur A M Wilde, Jean-Claude Tardif, Christopher J O'Donnell, Connie R Bezzina, Renu Virmani, Bruno H C H Stricker, Hanno L Tan, Christine M Albert, Aravinda Chakravarti, John D Rioux, Heikki V Huikuri, Sumeet S Chugh
ABSTRACT
Sudden cardiac death (SCD) continues to be one of the leading causes of mortality worldwide, with an annual incidence estimated at 250,000-300,000 in the United States and with the vast majority occurring in the setting of coronary disease. We performed a genome-wide association meta-analysis in 1,283 SCD cases and >20,000 control individuals of European ancestry from 5 studies, with follow-up genotyping in up to 3,119 SCD cases and 11,146 controls from 11 European ancestry studies, and identify the BAZ2B locus as associated with SCD (P = 1.8×10(-10)). The risk allele, while ancestral, has a frequency of ~1.4%, suggesting strong negative selection and increases risk for SCD by 1.92-fold per allele (95% CI 1.57-2.34). We also tested the role of 49 SNPs previously implicated in modulating electrocardiographic traits (QRS, QT, and RR intervals). Consistent with epidemiological studies showing increased risk of SCD with prolonged QRS/QT intervals, the interval-prolonging alleles are in aggregate associated with increased risk for SCD (P = 0.006).
Sigma-Aldrich
BAZ2B (2054-2168) human, recombinant, expressed in E. coli, ≥84% (SDS-PAGE)