Merck
  • Sleep Loss Can Cause Death through Accumulation of Reactive Oxygen Species in the Gut.
Sleep Loss Can Cause Death through Accumulation of Reactive Oxygen Species in the Gut.
Cell (2020-06-06)
Alexandra Vaccaro, Yosef Kaplan Dor, Keishi Nambara, Elizabeth A Pollina, Cindy Lin, Michael E Greenberg, Dragana Rogulja
ABSTRACT
The view that sleep is essential for survival is supported by the ubiquity of this behavior, the apparent existence of sleep-like states in the earliest animals, and the fact that severe sleep loss can be lethal. The cause of this lethality is unknown. Here we show, using flies and mice, that sleep deprivation leads to accumulation of reactive oxygen species (ROS) and consequent oxidative stress, specifically in the gut. ROS are not just correlates of sleep deprivation but drivers of death: their neutralization prevents oxidative stress and allows flies to have a normal lifespan with little to no sleep. The rescue can be achieved with oral antioxidant compounds or with gut-targeted transgenic expression of antioxidant enzymes. We conclude that death upon severe sleep restriction can be caused by oxidative stress, that the gut is central in this process, and that survival without sleep is possible when ROS accumulation is prevented. VIDEO ABSTRACT.
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide sodium salt
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium phenylbutyrate, ≥98% (HPLC)
Pricing and availability is not currently available.
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Quercetin, phyproof® Reference Substance
Pricing and availability is not currently available.
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Ebselen, cysteine modifier
Pricing and availability is not currently available.
Sigma-Aldrich
(±)-α-Lipoic acid, synthetic, ≥99% (titration), powder
Pricing and availability is not currently available.
Sigma-Aldrich
4-Hydroxy-TEMPO, 95%
Pricing and availability is not currently available.