Merck
  • Kinesin-7 CENP-E regulates chromosome alignment and genome stability of spermatogenic cells.
Kinesin-7 CENP-E regulates chromosome alignment and genome stability of spermatogenic cells.
Cell death discovery (2020-05-01)
Zhen-Yu She, Kai-Wei Yu, Ning Zhong, Yu Xiao, Ya-Lan Wei, Yang Lin, Yue-Ling Li, Ming-Hui Lu
ABSTRACT
Kinesin-7 CENP-E is an essential kinetochore motor required for chromosome alignment and congression. However, the specific functions of CENP-E in the spermatogenic cells during spermatogenesis remain unknown. In this study, we find that CENP-E proteins are expressed in the spermatogonia, spermatocytes, and the elongating spermatids. CENP-E inhibition by specific inhibitor GSK923295 results in the disruption of spermatogenesis and cell cycle arrest of spermatogenic cells. Both spermatogonia and spermatocytes are arrested in metaphase and several chromosomes are not aligned at the equatorial plate. We find that CENP-E inhibition leads to chromosome misalignment, the spindle disorganization, and the formation of the aneuploidy cells. Furthermore, the inhibition of CENP-E results in the defects in the formation of spermatids, including the sperm head condensation and the sperm tail formation. We have revealed that kinesin-7 CENP-E is essential for chromosome alignment and genome stability of the spermatogenic cells.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
GSK923295, ≥95% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human CENPE
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
L-(−)-Glucose, ≥99%