Merck
  • ZNF423 patient variants, truncations, and in-frame deletions in mice define an allele-dependent range of midline brain abnormalities.
ZNF423 patient variants, truncations, and in-frame deletions in mice define an allele-dependent range of midline brain abnormalities.
PLoS genetics (2020-09-15)
Ojas Deshpande, Raquel Z Lara, Oliver R Zhang, Dorothy Concepcion, Bruce A Hamilton
ABSTRACT
Interpreting rare variants remains a challenge in personal genomics, especially for disorders with several causal genes and for genes that cause multiple disorders. ZNF423 encodes a transcriptional regulatory protein that intersects several developmental pathways. ZNF423 has been implicated in rare neurodevelopmental disorders, consistent with midline brain defects in Zfp423-mutant mice, but pathogenic potential of most patient variants remains uncertain. We engineered ~50 patient-derived and small deletion variants into the highly-conserved mouse ortholog and examined neuroanatomical measures for 791 littermate pairs. Three substitutions previously asserted pathogenic appeared benign, while a fourth was effectively null. Heterozygous premature termination codon (PTC) variants showed mild haploabnormality, consistent with loss-of-function intolerance inferred from human population data. In-frame deletions of specific zinc fingers showed mild to moderate abnormalities, as did low-expression variants. These results affirm the need for functional validation of rare variants in biological context and demonstrate cost-effective modeling of neuroanatomical abnormalities in mice.
Sigma-Aldrich
Monoclonal Anti-Phosphoserine antibody produced in mouse, clone PSR-45, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Phosphothreonine antibody produced in mouse, clone PTR-8, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Phosphotyrosine antibody produced in mouse, clone PT-66, ascites fluid
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-ZNF423 Antibody, from rabbit, purified by affinity chromatography