Merck
  • Differential epigenetic regulation between the alternative promoters, PRDM1α and PRDM1β, of the tumour suppressor gene PRDM1 in human multiple myeloma cells.
Differential epigenetic regulation between the alternative promoters, PRDM1α and PRDM1β, of the tumour suppressor gene PRDM1 in human multiple myeloma cells.
Scientific reports (2020-09-29)
Raquel Romero-García, Laura Gómez-Jaramillo, Rosa María Mateos, Gema Jiménez-Gómez, Nuria Pedreño-Horrillo, Esther Foncubierta, Juan Francisco Rodríguez-Gutiérrez, Sebastián Garzón, Francisco Mora-López, Carmen Rodríguez, Luis M Valor, Antonio Campos-Caro
ABSTRACT
Multiple myeloma (MM) is a B-cell neoplasm that is characterized by the accumulation of malignant plasma cells in the bone marrow. The transcription factor PRDM1 is a master regulator of plasma cell development and is considered to be an oncosuppressor in several lymphoid neoplasms. The PRDM1β isoform is an alternative promoter of the PRDM1 gene that may interfere with the normal role of the PRDM1α isoform. To explain the induction of the PRDM1β isoform in MM and to offer potential therapeutic strategies to modulate its expression, we characterized the cis regulatory elements and epigenetic status of its promoter. We observed unexpected patterns of hypermethylation and hypomethylation at the PRDM1α and PRDM1β promoters, respectively, and prominent H3K4me1 and H3K9me2 enrichment at the PRDM1β promoter in non-expressing cell lines compared to PRDM1β-expressing cell lines. After treatment with drugs that inhibit DNA methylation, we were able to modify the activity of the PRDM1β promoter but not that of the PRDM1α promoter. Epigenetic drugs may offer the ability to control the expression of the PRDM1α/PRDM1β promoters as components of novel therapeutic approaches.
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone DO-1, purified from hybridoma cell culture