Merck
  • Single-cell transcriptome maps of myeloid blood cell lineages in Drosophila.
Single-cell transcriptome maps of myeloid blood cell lineages in Drosophila.
Nature communications (2020-09-10)
Bumsik Cho, Sang-Ho Yoon, Daewon Lee, Ferdinand Koranteng, Sudhir Gopal Tattikota, Nuri Cha, Mingyu Shin, Hobin Do, Yanhui Hu, Sue Young Oh, Daehan Lee, A Vipin Menon, Seok Jun Moon, Norbert Perrimon, Jin-Wu Nam, Jiwon Shim
ABSTRACT
The Drosophila lymph gland, the larval hematopoietic organ comprised of prohemocytes and mature hemocytes, has been a valuable model for understanding mechanisms underlying hematopoiesis and immunity. Three types of mature hemocytes have been characterized in the lymph gland: plasmatocytes, lamellocytes, and crystal cells, which are analogous to vertebrate myeloid cells, yet molecular underpinnings of the lymph gland hemocytes have been less investigated. Here, we use single-cell RNA sequencing to comprehensively analyze heterogeneity of developing hemocytes in the lymph gland, and discover previously undescribed hemocyte types including adipohemocytes, stem-like prohemocytes, and intermediate prohemocytes. Additionally, we identify the developmental trajectory of hemocytes during normal development as well as the emergence of the lamellocyte lineage following active cellular immunity caused by wasp infestation. Finally, we establish similarities and differences between embryonically derived- and larval lymph gland hemocytes. Altogether, our study provides detailed insights into the hemocyte development and cellular immune responses at single-cell resolution.
Sigma-Aldrich
Monoclonal Anti-Green Fluorescent Protein (GFP) antibody produced in mouse, clone GFP-20, ascites fluid
Glass beads, unwashed, 425-600 μm (30-40 U.S. sieve)