Merck
  • Epigenetic homogeneity in histone methylation underlies sperm programming for embryonic transcription.
Epigenetic homogeneity in histone methylation underlies sperm programming for embryonic transcription.
Nature communications (2020-07-15)
Mami Oikawa, Angela Simeone, Eva Hormanseder, Marta Teperek, Vincent Gaggioli, Alan O'Doherty, Emma Falk, Matthieu Sporniak, Clive D'Santos, Valar Nila Roamio Franklin, Kamal Kishore, Charles R Bradshaw, Declan Keane, Thomas Freour, Laurent David, Adrian T Grzybowski, Alexander J Ruthenburg, John Gurdon, Jerome Jullien
ABSTRACT
Sperm contributes genetic and epigenetic information to the embryo to efficiently support development. However, the mechanism underlying such developmental competence remains elusive. Here, we investigated whether all sperm cells have a common epigenetic configuration that primes transcriptional program for embryonic development. Using calibrated ChIP-seq, we show that remodelling of histones during spermiogenesis results in the retention of methylated histone H3 at the same genomic location in most sperm cell. This homogeneously methylated fraction of histone H3 in the sperm genome is maintained during early embryonic replication. Such methylated histone fraction resisting post-fertilisation reprogramming marks developmental genes whose expression is perturbed upon experimental reduction of histone methylation. A similar homogeneously methylated histone H3 fraction is detected in human sperm. Altogether, we uncover a conserved mechanism of paternal epigenetic information transmission to the embryo through the homogeneous retention of methylated histone in a sperm cells population.
Sigma-Aldrich
Anti-HMGB1 (HMG1) (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Cytochalasin B from Drechslera dematioidea, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder
Sigma-Aldrich
Anti-Histone H2A (acidic patch) Antibody, serum, Upstate®