Merck
  • Topologically unique heterometallic Cu(II)/Li coordination polymers self-assembled from N,N-bis(2-hydroxyethyl)-2-aminoethanesulfonic acid biobuffer: versatile catalyst precursors for mild hydrocarboxylation of alkanes to carboxylic acids.
Topologically unique heterometallic Cu(II)/Li coordination polymers self-assembled from N,N-bis(2-hydroxyethyl)-2-aminoethanesulfonic acid biobuffer: versatile catalyst precursors for mild hydrocarboxylation of alkanes to carboxylic acids.
Inorganic chemistry (2012-04-10)
Marina V Kirillova, Alexander M Kirillov, André N C Martins, Claudia Graiff, Antonio Tiripicchio, Armando J L Pombeiro
ABSTRACT
The facile aqueous medium reactions of copper(II) nitrate with BES biobuffer [(HOCH(2)CH(2))(2)N(CH(2)CH(2)SO(3)H), hereinafter referred as H(3)bes] in the presence of various benzenecarboxylic acids [benzoic (Hba), 3-hydroxybenzoic (Hhba), and 3,5-dihydroxybenzoic (Hdhba) acid] and lithium hydroxide gave rise to the self-assembly generation of three new heterometallic Cu(II)/Li materials, [Li(H(2)O)(4)][Cu(4)(μ(2)-Hbes)(4)(μ(2)-ba)]·H(2)O (1) and [Cu(4)(μ(3)-Hbes)(4)(L){Li(H(2)O)(2)}](n)·3nH(2)O {L = μ(2)-hba (2) and μ(2)-dhba (3)}. They were isolated as air-stable crystalline solids and fully characterized by infrared (IR) and UV-vis spectroscopy and electrospray ionization (ESI)-MS(±), elemental, thermal, and single-crystal X-ray diffraction analyses. The latter revealed that 1-3 have comparable packing patterns and unit cell parameters, being composed of similar [Cu(4)(μ-Hbes)(4)(μ-carboxylate)](-) cores and [Li(H(2)O)(4)](+) cations (in 1) or [μ-Li(H(2)O)(2)](+) groups (in 2 and 3), which are arranged into discrete 0D aggregates in 1 or infinite 3D noninterpenetrating metal-organic networks in 2 and 3. The topological analysis of the coordination polymers 2 and 3 disclosed the trinodal 3,3,4-connected underlying nets with an unprecedented topology defined by the point symbol of (4.6.8)(4)(4(2).6)(2)(6(2).16(2).18(2)), further simplification of which resulted in the binodal 4,4-connected nets with the pts (PtS) topology. Apart from representing very rare examples of coordination compounds derived from H(3)bes, 1-3 feature solubility in water and were applied as efficient and versatile catalyst precursors for the mild (60 °C) single-pot hydrocarboxylation, by CO and H(2)O, of various gaseous, linear, and cyclic C(n) (n = 2-9) alkanes into the corresponding C(n+1) carboxylic acids, in H(2)O/MeCN medium under homogeneous conditions and in the presence of potassium peroxodisulfate. Total yields (based on alkane) of carboxylic acids up to 78% were achieved, which are remarkable in the field of alkane functionalization under mild conditions, especially for a C-C bond formation reaction in aqueous acid-solvent-free medium.
Sigma-Aldrich
BES, ≥99.0% (titration)
Pricing and availability is not currently available.