Merck
  • Characterization of a neutral ceramidase orthologue from Aspergillus oryzae.
Characterization of a neutral ceramidase orthologue from Aspergillus oryzae.
FEMS microbiology letters (2009-08-05)
Sawaki Tada, Mayumi Matsushita-Morita, Satoshi Suzuki, Ken-Ichi Kusumoto, Yutaka Kashiwagi
ABSTRACT
Ceramide is an important molecule not only structurally but also regulationally as a modulator of various cellular events. Ceramidase (CDase) are classified into three different types (acid, alkaline, and neutral CDases). Neutral CDase could play an important role in the regulation of ceramide levels in the extracellular space. In this study, we describe the characterization of a neutral CDase orthologue from the filamentous fungus Aspergillus oryzae. The gene encoding the neutral CDase orthologue was cloned and overexpressed in A. oryzae. The purified recombinant enzyme was optimally active at pH 4.0-4.5 and 40 degrees C. The apparent K(m) and V(max) values of the enzyme for C12-NBD-ceramide were 3.32 microM and 0.085 micromol min(-1) mg(-1), respectively.
Product Description
Avanti
C12-NBD Sphingomyelin, Avanti Research - A Croda Brand 810219P, powder
Pricing and availability is not currently available.