Merck
  • Surfactant protein A (SP-A) binds to phosphatidylserine and competes with annexin V binding on late apoptotic cells.
Surfactant protein A (SP-A) binds to phosphatidylserine and competes with annexin V binding on late apoptotic cells.
Protein & cell (2011-01-05)
Anne Jäkel, Kenneth B M Reid, Howard Clark
ABSTRACT
The role of surfactant protein A (SP-A) in the recognition and clearance of apoptotic cells is well established, but to date, it is still not clear which surface molecules of apoptotic cells are involved in the process. Here we present evidence that phosphatidylserine (PS) is a relevant binding molecule for human SP-A. The binding is Ca(2+)-dependent and is not inhibited by mannose, suggesting that the sugar-binding site of the carbohydrate recognition domain (CRD) of SP-A is not involved. Flow cytometry studies on apoptotic Jurkat cells revealed apparent inhibition of annexin V binding by increasing concentrations of SP-A in late apoptotic but not early apoptotic cells, and this was consistent for Jurkat cells and neutrophils. Supporting these data, confocal microscopy results show a co-localisation of annexin V and SP-A in late apoptotic but not early apoptotic cells. However, we cannot conclude that this inhibition is exclusively due to the binding of SP-A to PS on the cell surface, as annexin V is not wholly specific for PS and SP-A also interacts with other phospholipids that might become exposed on the apoptotic cell surface.
Sigma-Aldrich
Streptavidin−Peroxidase from Streptomyces avidinii, lyophilized powder
Avanti
18:0-18:1 PS, 1-stearoyl-2-oleoyl-sn-glycero-3-phospho-L-serine (sodium salt), powder