Merck
Surfactant phospholipid metabolism.
Biochimica et biophysica acta (2012-10-03)
Marianna Agassandian, Rama K Mallampalli
ABSTRACT
Pulmonary surfactant is essential for life and is composed of a complex lipoprotein-like mixture that lines the inner surface of the lung to prevent alveolar collapse at the end of expiration. The molecular composition of surfactant depends on highly integrated and regulated processes involving its biosynthesis, remodeling, degradation, and intracellular trafficking. Despite its multicomponent composition, the study of surfactant phospholipid metabolism has focused on two predominant components, disaturated phosphatidylcholine that confers surface-tension lowering activities, and phosphatidylglycerol, recently implicated in innate immune defense. Future studies providing a better understanding of the molecular control and physiological relevance of minor surfactant lipid components are needed. This article is part of a Special Issue entitled Phospholipids and Phospholipid Metabolism.
Product Description
Avanti
14:0-12:0 NBD PG, Avanti Research - A Croda Brand 810162P, powder
Pricing and availability is not currently available.