Merck
Coenzyme A: back in action.
Progress in lipid research (2005-05-17)
Roberta Leonardi, Yong-Mei Zhang, Charles O Rock, Suzanne Jackowski
ABSTRACT
Coenzyme A (CoA) is a ubiquitous essential cofactor that plays a central role in the metabolism of carboxylic acids, including short- and long-chain fatty acids. In the last few years, all of the genes encoding the CoA biosynthetic enzymes have been identified and the structures of several proteins in the pathway have been determined. CoA is assembled in five steps from pantothenic acid and pathway intermediates are common to both prokaryotes and eukaryotes. In spite of the identical biochemistry, remarkable sequence differences among some of the prokaryotic and eukaryotic enzymes have been revealed by comparative genomics. Renewed interest in CoA has arisen from the realization that the biosynthetic pathway is a target for antibacterial drug discovery and from the unexpected association of a human neurodegenerative disorder with mutations in pantothenate kinase. The purpose of this review is to integrate previous knowledge with the most recent findings in the genetics, enzymology and regulation of CoA biosynthesis in bacteria, plants and mammals.
Product Description
Avanti
Coenzyme A (Li salt), Avanti Research - A Croda Brand 870701P, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Coenzyme A, oxidized lithium salt, ≥85%
Pricing and availability is not currently available.