Merck
Assembly and Folding Properties of Cytosolic IgG Intrabodies.
Scientific reports (2020-02-09)
Youngsil Seo, Yeonjin Lee, Minjae Kim, Hyunjoon Park, Myung-Hee Kwon
ABSTRACT
Intrabodies, antibodies expressed within cells, offer an interesting way to target intracellular molecules, making them potentially useful for biotechnology and medicine. However, it remains controversial whether full-size IgG intrabodies expressed in the reducing environment of the cytosol of mammalian cells are workable and structurally sound. Herein, we settle this issue with a systematic investigation of the structure and functionality of four chimeric IgG1s with distinct variable (V) domains but identical constant (C) domains. Full-size IgGs expressed in the cytosol of HEK293 cells were either assembly-competent or -incompetent, depending on the intrinsic properties of the V regions. Structural integrity of the C region is required for H:L association and the formation of a functional antigen-binding site. Partial intrachain disulfide bond formation occurs in both H and L chains of cytosolic IgG intrabodies, whereas interchain disulfide bond formation was absent and dispensable for functional assembly. IgG1s expressed in the cytosol and via the ER were shown to assemble differently. Our findings provide insight into the features and possible utilization of full-size IgGs as cytosolic antibodies in biotechnological and medical applications.
Sigma-Aldrich
IgG from human serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgM (μ-chain specific)−Agarose antibody produced in goat, IgG fraction of antiserum, saline suspension
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
RO-3306, ≥98% (HPLC)
Sigma-Aldrich
Anti-HA Tag Antibody, Upstate®, from mouse