Merck
  • Rigosertib-Activated JNK1/2 Eliminate Tumor Cells through p66Shc Activation.
Rigosertib-Activated JNK1/2 Eliminate Tumor Cells through p66Shc Activation.
Biology (2020-05-21)
Julia K Günther, Aleksandar Nikolajevic, Susanne Ebner, Jakob Troppmair, Sana Khalid
ABSTRACT
Rigosertib, via reactive oxygen species (ROS), stimulates cJun N-terminal kinases 1/2 (JNK1/2), which inactivate RAS/RAF signaling and thereby inhibit growth and survival of tumor cells. JNK1/2 are not only regulated by ROS-they in turn can also control ROS production. The prooxidant and cell death function of p66Shc requires phosphorylation by JNK1/2. Here, we provide evidence that establishes p66Shc, an oxidoreductase, as a JNK1/2 effector downstream of Rigosertib-induced ROS production, DNA damage, and cell death. This may provide a common pathway for suppression of tumor cell growth by Rigosertib.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Roche
Anti-HA-Peroxidase, High Affinity, from rat IgG1