Merck
  • Spaced Training Enhances Contextual Fear Memory via Activating Hippocampal 5-HT2A Receptors.
Spaced Training Enhances Contextual Fear Memory via Activating Hippocampal 5-HT2A Receptors.
Frontiers in molecular neuroscience (2020-02-11)
Lizhu Jiang, Liping Wang, Yan Yin, Mengke Huo, Chao Liu, Qixin Zhou, Dafu Yu, Lin Xu, Rongrong Mao
ABSTRACT
Spaced training is robustly superior to massed training, which is a well-documented phenomenon in humans and animals. However, the mechanisms underlying the spacing effect still remain unclear. We have reported previously that spacing training exerts memory-enhancing effects by inhibiting forgetting via decreasing hippocampal Rac1 activity. Here, using contextual fear conditioning in rat, we found that spaced but not massed training increased hippocampal 5-HT2A receptors' expression. Furthermore, hippocampal administration of 5-HT2A receptor antagonist MDL11939 before spaced training blocked the enhanced memory, while hippocampal administration of 5-HT2A receptor agonist TCB-2 before massed training promoted the memory. Moreover, MDL11939 activated hippocampal Rac1, while TCB-2 decreased hippocampal Rac1 activity in naïve rats. These results indicated the possibility of interaction between 5-HT2A receptors and Rac1, which was demonstrated by co-immunoprecipitation experiments. Our study first demonstrates that activation of hippocampal 5-HT2A is a mechanism underlying the spacing effect, and forgetting related molecular Rac1 is engaged in this process through interacting with 5-HT2A receptors, which suggest a promising strategy to modulate abnormal learning in cognitive disorders.
Sigma-Aldrich
Anti-Serotonin 2A (5-HT2A) Receptor (22-41) Rabbit pAb, liquid, Calbiochem®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Serotonin receptor 2A/HTR2A Antibody, clone 9B11.1, clone 9B11.1, from mouse
Pricing and availability is not currently available.