Merck
  • Translational derepression of Elavl4 isoforms at their alternative 5' UTRs determines neuronal development.
Translational derepression of Elavl4 isoforms at their alternative 5' UTRs determines neuronal development.
Nature communications (2020-04-05)
Tatiana Popovitchenko, Yongkyu Park, Nicholas F Page, Xiaobing Luo, Zeljka Krsnik, Yuan Liu, Iva Salamon, Jessica D Stephenson, Matthew L Kraushar, Nicole L Volk, Sejal M Patel, H R Sagara Wijeratne, Diana Li, Kandarp S Suthar, Aaron Wach, Miao Sun, Sebastian J Arnold, Wado Akamatsu, Hideyuki Okano, Luc Paillard, Huaye Zhang, Steven Buyske, Ivica Kostovic, Silvia De Rubeis, Ronald P Hart, Mladen-Roko Rasin
ABSTRACT
Neurodevelopment requires precise regulation of gene expression, including post-transcriptional regulatory events such as alternative splicing and mRNA translation. However, translational regulation of specific isoforms during neurodevelopment and the mechanisms behind it remain unknown. Using RNA-seq analysis of mouse neocortical polysomes, here we report translationally repressed and derepressed mRNA isoforms during neocortical neurogenesis whose orthologs include risk genes for neurodevelopmental disorders. We demonstrate that the translation of distinct mRNA isoforms of the RNA binding protein (RBP), Elavl4, in radial glia progenitors and early neurons depends on its alternative 5' UTRs. Furthermore, 5' UTR-driven Elavl4 isoform-specific translation depends on upstream control by another RBP, Celf1. Celf1 regulation of Elavl4 translation dictates development of glutamatergic neurons. Our findings reveal a dynamic interplay between distinct RBPs and alternative 5' UTRs in neuronal development and underscore the risk of post-transcriptional dysregulation in co-occurring neurodevelopmental disorders.
Sigma-Aldrich
5-Iodo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Anti-Nestin antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
5-Chloro-2′-deoxyuridine, thymidine analog
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-NeuN Antibody (rabbit), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit