- Application of immunoperoxidase techniques to localize horseradish peroxidase-tracer in the central nervous system.
Application of immunoperoxidase techniques to localize horseradish peroxidase-tracer in the central nervous system.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (1975-03-01)
L L Vacca, S L Rosario, E A Zimmerman, P Tomashefsky, N G Po-Ying, K C Hsu
PMID1127219
ABSTRACT
Immunoperoxidase techniques are presented which can be used to localize horseradish peroxidase-tracer in paraffin-embedded tissues of the central nervous system. Compared to histochemical methods using frozen sections, these immunologic techniques allow the use of stored, serial paraffin sections, and appear more sensitive for the demonstration of intraneuronal horseradish peroxidase after retrograde transport. The immunoperoxidase bridge techniques from reaction products of high quality which can easily be seen in fine processes.