Merck
  • Chemical Prelithiation of Negative Electrodes in Ambient Air for Advanced Lithium-Ion Batteries.
Chemical Prelithiation of Negative Electrodes in Ambient Air for Advanced Lithium-Ion Batteries.
ACS applied materials & interfaces (2019-02-20)
Gongwei Wang, Feifei Li, Dan Liu, Dong Zheng, Yang Luo, Deyu Qu, Tianyao Ding, Deyang Qu
ABSTRACT
This study reports an ambient-air-tolerant approach for negative electrode prelithiation by using 1 M lithium-biphenyl (Li-Bp)/tetrahydrofuran (THF) solution as the prelithiation reagent. Key to this strategy are the relatively stable nature of 1 M Li-Bp/THF in ambient air and the unique electrochemical behavior of Bp in ether and carbonate solvents. With its low redox potential of 0.41 V vs Li/Li+, Li-Bp can prelithiate various active materials with high efficacy. The successful prelithiation of a phosphrous/carbon composite electrode and the notable improvement in its initial Coulombic efficiency (CE) demonstrates the practicality of this strategy.
Sigma-Aldrich
1 M 9,9-dimethylfluorene in THF, a precursor solution to prepare the prelithiation reagent for Si anode
Pricing and availability is not currently available.
Sigma-Aldrich
1 M Biphenyl in DME, a precursor solution to prepare the prelithiation/presodiation reagent for disordered carbon, SiO, Sn, Sb, P, and S electrodes
Pricing and availability is not currently available.
Sigma-Aldrich
1 M Biphenyl in THF, a precursor solution to prepare the prelithiation/presodiation reagent for disordered carbon, SiO, Sn, Sb, P, and S electrodes
Pricing and availability is not currently available.