Merck
  • Metabolomic Signature in Sera of Multiple Sclerosis Patients during Pregnancy.
Metabolomic Signature in Sera of Multiple Sclerosis Patients during Pregnancy.
International journal of molecular sciences (2018-11-18)
Claudia Rossi, Ilaria Cicalini, Mirco Zucchelli, Maria di Ioia, Marco Onofrj, Luca Federici, Piero Del Boccio, Damiana Pieragostino
ABSTRACT
Multiple sclerosis (MuS) is an autoimmune disease of the central nervous system characterized by neuroinflammation, neurodegeneration, and degradation of the myelin sheath. Epidemiological studies have shown that the female gender is more susceptible than the male gender to MuS development, with a female-to-male ratio of 2:1. Despite this high onset, women have a better prognosis than men, and the frequency of the relapsing phase decreases during pregnancy, while it increases soon after birth. Therefore, it is interesting to investigate hormonal fluctuations during pregnancy and whether they correlate with metabolic signatures. To gain a deeper inside into the biochemical mechanism of such a multifactorial disease, we adopted targeted metabolomics approaches for the determination of many serum metabolites in 12 pregnant women affected by MuS by mass spectrometry analysis. Our data show a characteristic hormonal fluctuation for estrogens and progesterone, as expected. They also highlight other interesting hormonal alterations for cortisol, corticosterone, 11-deoxycortisol, 4-androstene-3,17-dione, testosterone, and 17α-hydroxyprogesterone. Furthermore, a negative correlation with progesterone levels was observed for amino acids and for acylcarnitines, while an imbalance of different sphingolipids pathways was found during pregnancy. In conclusion, these data are in agreement with the characteristic clinical signs of MuS patients during pregnancy and, if confirmed, they may add an important tessera in the complex mosaic of maternal neuroprotection.
Product Description
Avanti
17:0-20:4 PI(3,4,5)P3, Avanti Research - A Croda Brand LM1906, powder
Pricing and availability is not currently available.
Avanti
Sphingosine (d17:1), Avanti Research - A Croda Brand LM2000, ethanol solution
Pricing and availability is not currently available.