Merck
  • Monitoring cell endocytosis of liposomes by real-time electrical impedance spectroscopy.
Monitoring cell endocytosis of liposomes by real-time electrical impedance spectroscopy.
Analytical and bioanalytical chemistry (2020-05-27)
Claudia Caviglia, Francesca Garbarino, Chiara Canali, Fredrik Melander, Roberto Raiteri, Giorgio Ferrari, Marco Sampietro, Arto Heiskanen, Thomas Lars Andresen, Kinga Zór, Jenny Emnéus
ABSTRACT
Evaluation and understanding the effect of drug delivery in in vitro systems is fundamental in drug discovery. We present an assay based on real-time electrical impedance spectroscopy (EIS) measurements that can be used to follow the internalisation and cytotoxic effect of a matrix metalloproteinase (MMP)-sensitive liposome formulation loaded with oxaliplatin (OxPt) on colorectal cancer cells. The EIS response identified two different cellular processes: (i) a negative peak in the cell index (CI) within the first 5 h, due to onset of liposome endocytosis, followed by (ii) a subsequent CI increase, due to the reattachment of cells until the onset of cytotoxicity with a decrease in CI. Free OxPt or OxPt-loaded Stealth liposomes did not show this two-stage EIS response; the latter can be due to the fact that Stealth cannot be cleaved by MMPs and thus is not taken up by the cells. Real-time bright-field imaging supported the EIS data, showing variations in cell adherence and cell morphology after exposure to the different liposome formulations. A drastic decrease in cell coverage as well as rounding up of cells during the first 5 h of exposure to OxPt-loaded (MMP)-sensitive liposome formulation is reflected by the first negative EIS response, which indicates the onset of liposome endocytosis. Graphical abstract.
Product Description
Avanti
DSPE-PEG(2000) Amine, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
18:0 PC (DSPC), Avanti Research - A Croda Brand
Avanti
DSPE-PEG(2000) Amine, Avanti Research - A Croda Brand 880128C
Avanti
18:1 TAP (DOTAP), Avanti Research - A Croda Brand