Merck
  • Role of TNFα and leptin signaling in colon cancer incidence and tumor growth under obese phenotype.
Role of TNFα and leptin signaling in colon cancer incidence and tumor growth under obese phenotype.
Biochimica et biophysica acta. Molecular basis of disease (2020-01-01)
Snahlata Singh, Shyamananda Singh Mayengbam, Surbhi Chouhan, Bhavana Deshmukh, Pranay Ramteke, Dipti Athavale, Manoj Kumar Bhat
ABSTRACT
Epidemiological studies over the last few decades have shown a strong influence of obesity on colon cancer risk and its progression. These studies have primarily focussed on the role of adipokines in driving cancer progression. We investigated the incidence of cancerous polyp formation and tumor progression in presence and absence of functional leptin along with exploring the role of tumor necrosis factor α (TNFα), under obese condition. By utilizing diet induced obese and genetically obese mice, carcinogen induced colon polyp formation was investigated. Experiments were performed using tumor tissues and cell lines to delineate the inter-relationship between leptin and TNFα. Data shown in this report indicates that in leptin knockdown obese mice, AOM/DSS induced polyps are smaller and lesser in numbers as compared to AOM/DSS induced polyps in diet induced obese mice. Further in vitro experiments suggest that abrogation of leptin associated pathways promote TNFα induced apoptosis. Mechanistically, we report that TNFα induces p53 independent cell death through up regulation of p53 upregulated modulator of apoptosis (PUMA). TNFα induced PUMA was inhibited upon pre- exposure of cells to leptin, prior to TNFα treatment. Collectively these results indicate that obesity due to leptin non-functionality facilitates TNFα induced colon cancer cell death.
Sigma-Aldrich
Leptin human, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Bay 11-7082, ≥98% (HPLC), powder
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, ≥98% (HPLC)
Sigma-Aldrich
Azoxymethane, 13.4 M, ≥98%