Merck
  • Targeted degradation of anaplastic lymphoma kinase by gold nanoparticle-based multi-headed proteolysis targeting chimeras.
Targeted degradation of anaplastic lymphoma kinase by gold nanoparticle-based multi-headed proteolysis targeting chimeras.
Colloids and surfaces. B, Biointerfaces (2020-01-29)
Yingming Wang, Lingfei Han, Fulei Liu, Fubai Yang, Xueyang Jiang, Haopeng Sun, Feng Feng, Jingwei Xue, Wenyuan Liu
ABSTRACT
Anaplastic lymphoma kinase (ALK) is a major target in treating non-small-cell lung cancer, and several ALK inhibitors have been developed to antagonize its kinase activity. However, patients treated with inhibitors ultimately develop drug resistance. Therefore, therapies with new mechanisms of action are needed. Proteolysis targeting chimeras (PROTACs) are molecules that comprise a ligand for binding a protein of interest (POI), a connecting linker and a ligand for recruiting E3 ligase, and cause degradation of the target POI. Here, the first multi-headed PROTAC, as a proof of concept, is developed as a gold nanoparticle (GNP)-based drug delivery system for delivering PROTACs to target ALK. Pegylated GNPs loaded with both ceritinib and pomalidomide molecules, termed Cer/Pom-PEG@GNPs, showed good stability in several media. The GNP conjugates potently decreased the levels of ALK fusion proteins in a dose- and time-dependent manner, and specifically inhibited the proliferation of NCI-H2228 cells. In comparison with small molecule PROTACs, the new multi-headed PROTAC promoted the formation of coacervates of POIs/multi-headed PROTAC/E3 ubiquitin ligases, and POI and E3 ubiquitin ligase interacted through multidirectional ligands and a flexible linker, thereby avoiding the need for complicated structure optimization of PROTACs. In conclusion, Cer/Pom-PEG@GNPs can degrade intracellular ALK fusion proteins with minor off-target toxicity and can be applied in patients resistant to ALK inhibitors. As a nano-based drug carrier, Cer/Pom-PEG@GNPs have the potential to enable prolonged circulation and specifically distribute drugs to tumor regions in vivo; thus, further investigation is warranted.
Sigma-Aldrich
3-Fluorophthalic anhydride, 95%
Pricing and availability is not currently available.