Merck
  • Effects of two little-studied environmental pollutants on early development in anurans.
Effects of two little-studied environmental pollutants on early development in anurans.
Environmental pollution (Barking, Essex : 1987) (2020-02-12)
Veronika Bókony, Viktória Verebélyi, Nikolett Ujhegyi, Zsanett Mikó, Edina Nemesházi, Márk Szederkényi, Stephanie Orf, Evelin Vitányi, Ágnes M Móricz
ABSTRACT
Despite intensive ecotoxicological research, we still know relatively little about the ecological impacts of many environmental contaminants. Filling these knowledge gaps is particularly important regarding amphibians, because they play significant roles in freshwater and terrestrial ecosystems, and their populations are declining worldwide. In this study, we investigated two pollutants that have been poorly studied in ecotoxicology despite their widespread occurrence in surface waters: the herbicide terbuthylazine and the pharmaceutical drug carbamazepine. We exposed two anuran species throughout their larval development to each of two environmentally relevant concentrations of each pollutant, and recorded mortality and 17 sub-lethal endpoints up to several months after exposure. Mortality was low and unrelated to treatment. In agile frogs (Rana dalmatina), we found that treatment with 0.3 μg/L terbuthylazine decreased tadpole activity and reduced fat bodies in juveniles, whereas treatment with 50 μg/L carbamazepine decreased spleen size and increased spleen pigmentation. In common toads (Bufo bufo), treatment with 0.003 μg/L terbuthylazine increased body mass at metamorphosis, treatment with 0.3 μg/L terbuthylazine increased the size of optic tecta, and treatment with 0.5 μg/L carbamazepine decreased hypothalamus size. Treatment with 50 μg/L carbamazepine reduced the feeding activity of toad tadpoles, decreased their production of anti-predatory bufadienolide toxins, and increased their body mass at metamorphosis; juvenile toads in this treatment group had reduced spleen pigmentation. Neither treatments affected the time to metamorphosis, post-metamorphic body mass, or sex ratios significantly. These results show that environmental levels of both terbuthylazine and carbamazepine can have several sub-lethal effects on anurans, which may be detrimental to individual fitness and population persistence in natural conditions. Our findings further highlight that toxic effects cannot be generalized between chemicals of similar structure, because the terbuthylazine effects we found do not conform with previously reported effects of atrazine, a related and extensively studied herbicide.
Product Description
Supelco
Terbuthylazine, PESTANAL®, analytical standard