Merck
  • A Conserved Mito-Cytosolic Translational Balance Links Two Longevity Pathways.
A Conserved Mito-Cytosolic Translational Balance Links Two Longevity Pathways.
Cell metabolism (2020-02-23)
Marte Molenaars, Georges E Janssens, Evan G Williams, Aldo Jongejan, Jiayi Lan, Sylvie Rabot, Fatima Joly, Perry D Moerland, Bauke V Schomakers, Marco Lezzerini, Yasmine J Liu, Mark A McCormick, Brian K Kennedy, Michel van Weeghel, Antoine H C van Kampen, Ruedi Aebersold, Alyson W MacInnes, Riekelt H Houtkooper
ABSTRACT
Slowing down translation in either the cytosol or the mitochondria is a conserved longevity mechanism. Here, we found a non-interventional natural correlation of mitochondrial and cytosolic ribosomal proteins (RPs) in mouse population genetics, suggesting a translational balance. Inhibiting mitochondrial translation in C. elegans through mrps-5 RNAi repressed cytosolic translation. Transcriptomics integrated with proteomics revealed that this inhibition specifically reduced translational efficiency of mRNAs required in growth pathways while increasing stress response mRNAs. The repression of cytosolic translation and extension of lifespan from mrps-5 RNAi were dependent on atf-5/ATF4 and independent from metabolic phenotypes. We found the translational balance to be conserved in mammalian cells upon inhibiting mitochondrial translation pharmacologically with doxycycline. Lastly, extending this in vivo, doxycycline repressed cytosolic translation in the livers of germ-free mice. These data demonstrate that inhibiting mitochondrial translation initiates an atf-5/ATF4-dependent cascade leading to coordinated repression of cytosolic translation, which could be targeted to promote longevity.
Sigma-Aldrich
Doxycycline hyclate
Pricing and availability is not currently available.
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Pricing and availability is not currently available.
Supelco
Coated GC Packing, phase SP®-1000, 1%, matrix 60/80 Carbopack B support, bottle of 15 g
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Pricing and availability is not currently available.