Merck
  • Pluripotency factor NANOG promotes germ cell maintenance in vitro without triggering dedifferentiation of spermatogonial stem cells.
Pluripotency factor NANOG promotes germ cell maintenance in vitro without triggering dedifferentiation of spermatogonial stem cells.
Theriogenology (2020-03-08)
Mi Yang, Bingquan Deng, Lei Geng, Lufan Li, Xin Wu
ABSTRACT
Spermatogonial stem cells (SSCs) are defined as the tissue-specific stem cells in the testes that produce sperm and support life-long spermatogenesis. A culture microenvironment could convert mouse SSCs from unipotent to pluripotent states; however, the underlying mechanism is unclear. NANOG has been considered the decisive transcriptional factor for pluripotency transition of stem cells, but NANOG is not expressed in SSCs. Here, we investigated whether NANOG overexpression could result in SSCs being converted into a pluripotent state. We found that rare NANOG-positive cells could be detected in spermatogonia, pachytene spermatocytes, and even round spermatids in mouse testes, and that the induction of NANOG could promote the proliferation of cultured SSCs in vitro and partially compensate for the role of the growth factor GDNF. In vivo allogeneic transplantation of NANOG-overexpressing germ cells did not yield any teratoma-like tissues, but regenerated normal colonies of spermatogenesis in the testes of recipient mice. Collectively, our data showed that overexpression of the pluripotency factor NANOG along did not dedifferentiate testis germline stem cells into a pluripotent state, suggesting that other genetic or epigenetic factors are involved in SSC reprogramming.
Product Description
Supelco
Palmitoleic acid, analytical standard
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Linolenic acid, ≥99%
Sigma-Aldrich
holo-Transferrin bovine, Iron-saturated, BioReagent, suitable for cell culture
Sigma-Aldrich
Putrescine dihydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Insulin from bovine pancreas, ≥25 USP units/mg (HPLC), powder
Sigma-Aldrich
Palmitoleic acid, ≥98.5% (GC), liquid
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside, ≥98%, powder
Supelco
Busulfan, analytical standard, for drug analysis
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Hartman′s Fixative, histological tissue fixative