MilliporeSigma
Two-Dimensional van der Waals Nanoplatelets with Robust Ferromagnetism.
Nano letters (2020-02-08)
Michael C De Siena, Sidney E Creutz, Annie Regan, Paul Malinowski, Qianni Jiang, Kyle T Kluherz, Guomin Zhu, Zhong Lin, James J De Yoreo, Xiaodong Xu, Jiun-Haw Chu, Daniel R Gamelin
ABSTRACT
We have synthesized unique colloidal nanoplatelets of the two-dimensional (2D) van der Waals ferromagnet CrI3 and have characterized these nanoplatelets structurally, magnetically, and by magnetic circular dichroism spectroscopy. The CrI3 nanoplatelets have lateral dimensions of ∼25 nm and thicknesses of only ∼4 nm, corresponding to just a few CrI3 monolayers. Magnetic and magneto-optical measurements demonstrate robust 2D ferromagnetic ordering with Curie temperatures similar to bulk CrI3, despite their small size. These data also show magnetization steps akin to those observed in micron-sized few-layer 2D sheets associated with concerted spin-reversal of individual CrI3 layers within few-layer van der Waals stacks. Similar data have also been obtained for CrBr3 and anion-alloyed Cr(I1-xBr x )3 nanoplatelets. These results represent the first example of lateral nanostructures of 2D van der Waals ferromagnets of any composition. The demonstration of robust ferromagnetism at nanometer lateral dimensions opens new doors for miniaturization in spintronics devices based on van der Waals ferromagnets.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Iodotrimethylsilane, 97%