Merck
  • Reciprocal cross-sensitization of D1 and D3 receptors following pharmacological stimulation in the hemiparkinsonian rat.
Reciprocal cross-sensitization of D1 and D3 receptors following pharmacological stimulation in the hemiparkinsonian rat.
Psychopharmacology (2019-08-23)
Kathryn Lanza, Katherine Chemakin, Sarah Lefkowitz, Carolyn Saito, Nicole Chambers, Christopher Bishop
ABSTRACT
In the majority of Parkinson's disease (PD) patients, long-term dopamine (DA) replacement therapy leads to dyskinesia characterized by abnormal involuntary movements (AIMs). There are various mechanisms of dyskinesia, such as the sensitization of striatal DA D1 receptors (D1R) and upregulation of DA D3 receptors (D3R). These receptors interact physically and functionally in D1R-bearing medium spiny neurons to synergistically drive dyskinesia. However, the cross-receptor-mediated effects due to D1R-D3R cooperativity are still poorly understood. In pursuit of this, we examined whether or not pharmacological D1R or D3R stimulation sensitizes the dyskinetic response to the appositional agonist, a process known as cross-sensitization. First, we established D1R-D3R behavioral synergy in a cohort of 6-OHDA-lesioned female adult Sprague-Dawley rats. Then, in a new cohort, we tested for cross-sensitization in a between-subject design. Five groups received a sub-chronic regimen of either saline, the D1R agonist SKF38393 (1.0 mg/kg), or the D3R agonist PD128907 (0.3 mg/kg). For the final injection, each group received an acute injection of the other agonist. AIMs were monitored following each injection. Sub-chronic administration of both SKF38393 and PD128907 induced the development of dyskinesia. More importantly, cross-agonism tests revealed reciprocal cross-sensitization; chronic treatment with either SKF38393 or PD128907 induced sensitization to a single administration of the other agonist. This reciprocity was not marked by changes to either D1R or D3R striatal mRNA expression. The current study provides key behavioral data demonstrating the role of D3R in dyskinesia and provides behavioral evidence of D1R and D3R functional interactions.
Sigma-Aldrich
Hydrazine monohydrochloride, 97%