Merck
  • Synthesis of Ultrahigh Molecular Weight Polymers Containing Reactive Functionality with Low PDIs by Polymerizations of Long-Chain α-Olefins in the Presence of Their Nonconjugated Dienes by Cp*TiMe2(O-2,6-iPr2C6H3)-Borate Catalyst.
Synthesis of Ultrahigh Molecular Weight Polymers Containing Reactive Functionality with Low PDIs by Polymerizations of Long-Chain α-Olefins in the Presence of Their Nonconjugated Dienes by Cp*TiMe2(O-2,6-iPr2C6H3)-Borate Catalyst.
Polymers (2019-12-22)
Kotohiro Nomura, Sarntamon Pengoubol, Wannida Apisuk
ABSTRACT
Copolymerizations of 1-decene (DC) with 1,9-decadiene (DCD), 1-dodecene (DD) with 1,11-dodecadiene (DDD), and 1-tetradecene (TD) with 1,13-tetradecadiene (TDD), using Cp*TiMe2(O-2,6-iPr2C6H3) (1)-[Ph3C][B(C6F5)4] (borate) catalyst in the presence of AliBu3/Al(n-C8H17)3 proceeded in a quasi-living manner in n-hexane at -30 to -50 °C, affording ultrahigh molecular weight (UHMW) copolymers containing terminal olefinic double bonds in the side chain with rather low PDI (Mw/Mn) values. In the DC/DCD copolymerization, the resultant copolymer prepared at -40 °C possessed UHMW (Mn = 1.40 × 106 after 45 min) with low PDI (Mw/Mn = 1.39); both the activity and the PDI value decreased at low polymerization temperature (Mn = 5.38 × 105, Mw/Mn = 1.18, after 120 min at -50 °C). UHMW poly(TD-co-TDD) was also obtained in the copolymerization at -30 °C (Mn = 9.12 × 105, Mw/Mn = 1.51, after 120 min), using this catalyst.
Sigma-Aldrich
1,9-Decadiene, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
1,13-Tetradecadiene, 90%
Pricing and availability is not currently available.