Merck
  • Development of a monoclonal antibody-based immunochromatographic assay for the detection of carbamazepine and carbamazepine-10, 11-epoxide.
Development of a monoclonal antibody-based immunochromatographic assay for the detection of carbamazepine and carbamazepine-10, 11-epoxide.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2020-02-29)
Shengyang Zhou, Liguang Xu, Liqiang Liu, Hua Kuang, Chuanlai Xu
ABSTRACT
In this study, haptens were designed to produce highly sensitive and specific monoclonal antibodies (mAb) against carbamazepine (CBZ) and its metabolite carbamazepine-10, 11-epoxide (CBZ-EP). According to the results of our competitive enzyme-linked immunosorbent assay (ic-ELISA), the half-maximum inhibitory concentration values for anti-CBZ and anti-CBZ-EP mAb were 0.18 and 0.59 ng/mL, respectively. An immunochromatographic assay (ICA) was developed for the determination of CBZ and CBZ-EP concentrations. This method can provide visible limits of detection ranging from 0.25 to 1 ng/mL, and cut-off limits ranging from 5 to 10 ng/mL, and takes 10 min to evaluate with the naked eye. Importantly, these observations were consistent with those obtained by ic-ELISA and liquid chromatography-mass spectrometry. The ICA assay represented a reliable, fast, and high-throughput method for the determination of CBZ and CBZ-EP in serum samples.