Merck
  • Intercalating pyrene with polypeptide as a novel self-assembly nano-carrier for colon cancer suppression in vitro and in vivo.
Intercalating pyrene with polypeptide as a novel self-assembly nano-carrier for colon cancer suppression in vitro and in vivo.
Materials science & engineering. C, Materials for biological applications (2020-04-02)
Pei-Ying Lo, Guang-Yu Lee, Jia-Huei Zheng, Jen-Hsien Huang, Er-Chieh Cho, Kuen-Chan Lee
ABSTRACT
Giving patients right dosage is an essential concept of precision medicine. Most of nanocarriers lack of flexible drug capacity and structural stability to be customized for specific treatment, resulting in low therapeutic efficacy and unexpected side effects. Thus, a growing need emerges for fast and rigorous approaches to develop nanoparticles with properties of adjustable dosage and controllable particle size. Poly-l-Lysine is known for its enhanced bioadhesivity and pH-triggered structural swelling effect, which is utilized as the main agent to activate the multistage drug releasing. Inspired by natural bio-assembly system, we report a simple method to self-assemble Poly-l-Lysine-based nanoparticles via supramolecular recognitions of cross-linked pyrenes, which provides noncovalent force to flexibly encapsulate Doxorubincin and to construct robust nanostructures. Pyrene-modified polypeptide self-assemblies are able to adjust drug payload from 1: 10 to 2:1 (drug: polypeptide) without changing its uniform nano-spherical morphology. This nanostructure remained the as-made morphology even after experiencing the long-term (~ 10 weeks) storage at room temperature. Also, the nanoparticles displayed multi-step drug release behaviours and exhibited great in vitro and in vivo cytotoxicity towards colon cancer cells. The as-mentioned nanoparticles provide a novel perspective to compensate the clinical needs of specific drug feedings and scalable synthesis with advantages of simple-synthesis, size-adaptivity, and morphology reversibility.
Sigma-Aldrich
1-Pyrenebutyric acid N-hydroxysuccinimide ester, 95%
Pricing and availability is not currently available.