Merck
  • H1R mediates local anesthetic-induced vascular permeability in angioedema.
H1R mediates local anesthetic-induced vascular permeability in angioedema.
Toxicology and applied pharmacology (2020-02-18)
Jiao Cao, Yongjing Zhang, Delu Che, Rui Liu, Liu Yang, Tao Zhang, Langchong He
ABSTRACT
Angioedema may occur during local anesthetic (LA) injection in the perioperative period. Histaminergic angioedema is the most common form of angioedema. It has been reported that LA is a potential exogenous ligand for histamine receptor 1 (H1R). Whether H1R participates in LA-induced angioedema is still controversial. By using a constructed H1R high-expressed cell model, siRNA transfection, pharmacologic means, and genetically modified animal models, here we showed that H1R mediated LA-induced hyperpermeability. LA with uncycled N-methyl scaffold in the side chain (procaine, tetracaine and lidocaine) had a better strength of drug-H1R affinity than that for LA with cycled N atom (bupivacaine and ropivacaine) by the molecular docking assay and equilibrium dissociation constant (KD values) obtained from the cell membrane chromatography (CMC) relative standard method. Procaine, tetracaine, and lidocaine triggered big calcium mobilization in H1R-HEK293 cells and human umbilical vein endothelial cells (HUVECs) but much weaker in NC-HEK293 cells or H1R knockdown HUVECs. Besides, the results of transendothelial resistance measurement, paracellular flux assay and immunofluorescence showed that procaine induced H1R-dependent hyperpermeability, which involved in PLCγ/IP3R/PKC, ERK1/2, Akt signaling pathways, downstream vascular endothelial cadherin (VE-cad) destabilization. Furthermore, H1R gene knockout prevented paw swelling and vascular leakage caused by procaine, tetracaine, and lidocaine in vivo. This study supported a key role of H1R in LA-induced angioedema, and suggested that in the design of LA structure, the ring formation of the N-methyl scaffold on the side chain can properly avoid the angioedema.
Sigma-Aldrich
MISSION® esiRNA, targeting human HRH1
Sigma-Aldrich
Evans Blue, Dye content ≥75 %
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%
Sigma-Aldrich
Dimethyl sulfoxide