Merck
  • Characterization of the highly active polyhydroxyalkanoate synthase of Chromobacterium sp. strain USM2.
Characterization of the highly active polyhydroxyalkanoate synthase of Chromobacterium sp. strain USM2.
Applied and environmental microbiology (2011-03-15)
Kesaven Bhubalan, Jo-Ann Chuah, Fumi Shozui, Christopher J Brigham, Seiichi Taguchi, Anthony J Sinskey, Chokyun Rha, Kumar Sudesh
ABSTRACT
The synthesis of bacterial polyhydroxyalkanoates (PHA) is very much dependent on the expression and activity of a key enzyme, PHA synthase (PhaC). Many efforts are being pursued to enhance the activity and broaden the substrate specificity of PhaC. Here, we report the identification of a highly active wild-type PhaC belonging to the recently isolated Chromobacterium sp. USM2 (PhaC(Cs)). PhaC(Cs) showed the ability to utilize 3-hydroxybutyrate (3HB), 3-hydroxyvalerate (3HV), and 3-hydroxyhexanoate (3HHx) monomers in PHA biosynthesis. An in vitro assay of recombinant PhaC(Cs) expressed in Escherichia coli showed that its polymerization of 3-hydroxybutyryl-coenzyme A activity was nearly 8-fold higher (2,462 ± 80 U/g) than that of the synthase from the model strain C. necator (307 ± 24 U/g). Specific activity using a Strep2-tagged, purified PhaC(Cs) was 238 ± 98 U/mg, almost 5-fold higher than findings of previous studies using purified PhaC from C. necator. Efficient poly(3-hydroxybutyrate) [P(3HB)] accumulation in Escherichia coli expressing PhaC(Cs) of up to 76 ± 2 weight percent was observed within 24 h of cultivation. To date, this is the highest activity reported for a purified PHA synthase. PhaC(Cs) is a naturally occurring, highly active PHA synthase with superior polymerizing ability.
Sigma-Aldrich
DL-β-Hydroxybutyryl coenzyme A lithium salt, ≥90%
Pricing and availability is not currently available.