Merck
  • Ebselen as template for stabilization of A4V mutant dimer for motor neuron disease therapy.
Ebselen as template for stabilization of A4V mutant dimer for motor neuron disease therapy.
Communications biology (2020-03-07)
Varunya Chantadul, Gareth S A Wright, Kangsa Amporndanai, Munazza Shahid, Svetlana V Antonyuk, Gina Washbourn, Michael Rogers, Natalie Roberts, Matthew Pye, Paul M O'Neill, S Samar Hasnain
ABSTRACT
Mutations to the gene encoding superoxide dismutase-1 (SOD1) were the first genetic elements discovered that cause motor neuron disease (MND). These mutations result in compromised SOD1 dimer stability, with one of the severest and most common mutations Ala4Val (A4V) displaying a propensity to monomerise and aggregate leading to neuronal death. We show that the clinically used ebselen and related analogues promote thermal stability of A4V SOD1 when binding to Cys111 only. We have developed a A4V SOD1 differential scanning fluorescence-based assay on a C6S mutation background that is effective in assessing suitability of compounds. Crystallographic data show that the selenium atom of these compounds binds covalently to A4V SOD1 at Cys111 at the dimer interface, resulting in stabilisation. This together with chemical amenability for hit expansion of ebselen and its on-target SOD1 pharmacological chaperone activity holds remarkable promise for structure-based therapeutics for MND using ebselen as a template.
Sigma-Aldrich
cis-11-Methyl-2-dodecenoic acid, ≥90.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
MR6-31-2, ≥98% (HPLC)
Pricing and availability is not currently available.