Merck
  • Paracetamol is a centrally acting analgesic using mechanisms located in the periaqueductal grey.
Paracetamol is a centrally acting analgesic using mechanisms located in the periaqueductal grey.
British journal of pharmacology (2019-11-18)
David André Barrière, Fawzi Boumezbeur, Romain Dalmann, Roberto Cadeddu, Damien Richard, Jérémy Pinguet, Laurence Daulhac, Philippe Sarret, Kevin Whittingstall, Matthieu Keller, Sébastien Mériaux, Alain Eschalier, Christophe Mallet
ABSTRACT
We previously demonstrated that paracetamol has to be metabolised in the brain by fatty acid amide hydrolase enzyme into AM404 (N-(4-hydroxyphenyl)-5Z,8Z,11Z,14Z-eicosatetraenamide) to activate CB1 receptors and TRPV1 channels, which mediate its analgesic effect. However, the brain mechanisms supporting paracetamol-induced analgesia remain unknown. The effects of paracetamol on brain function in Sprague-Dawley rats were determined by functional MRI. Levels of neurotransmitters in the periaqueductal grey (PAG) were measured using in vivo 1 H-NMR and microdialysis. Analgesic effects of paracetamol were assessed by behavioural tests and challenged with different inhibitors, administered systemically or microinjected in the PAG. Paracetamol decreased the connectivity of major brain structures involved in pain processing (insula, somatosensory cortex, amygdala, hypothalamus, and the PAG). This effect was particularly prominent in the PAG, where paracetamol, after conversion to AM404, (a) modulated neuronal activity and functional connectivity, (b) promoted GABA and glutamate release, and (c) activated a TRPV1 channel-mGlu5 receptor-PLC-DAGL-CB1 receptor signalling cascade to exert its analgesic effects. The elucidation of the mechanism of action of paracetamol as an analgesic paves the way for pharmacological innovations to improve the pharmacopoeia of analgesic agents.
Sigma-Aldrich
Monoclonal Anti-FAAH antibody produced in mouse, clone 4H8, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Capsazepine, ≥98% (HPLC), solid