Merck
  • Increased Levels of Plasma Tumor Necrosis Factor-α Mediate Schizophrenia Symptom Dimensions and Neurocognitive Impairments and Are Inversely Associated with Natural IgM Directed to Malondialdehyde and Paraoxonase 1 Activity.
Increased Levels of Plasma Tumor Necrosis Factor-α Mediate Schizophrenia Symptom Dimensions and Neurocognitive Impairments and Are Inversely Associated with Natural IgM Directed to Malondialdehyde and Paraoxonase 1 Activity.
Molecular neurobiology (2020-02-11)
Michael Maes, Sunee Sirivichayakul, Andressa Keiko Matsumoto, Annabel Maes, Ana Paula Michelin, Laura de Oliveira Semeão, João Victor de Lima Pedrão, Estefania G Moreira, Decio S Barbosa, Michel Geffard, Andre F Carvalho, Buranee Kanchanatawan
ABSTRACT
Accumulating evidence suggests that TNF-α-mediated immune-neurotoxicity contributes to cognitive impairments and the overall severity of schizophrenia (OSOS). There are no data whether peripheral IL-6 and IL-4 may affect the phenome of schizophrenia above and beyond the effects of TNF-α and whether those cytokines are regulated by lowered natural IgM to malondialdehyde (MDA) and paraoxonase 1 enzyme activity. We assessed the aforementioned biomarkers in a cross-sectional study that enrolled schizophrenia patients with (n = 40) and without (n = 40) deficit schizophrenia and 40 healthy controls. Deficit schizophrenia was best predicted by a combination of increased IL-6 and PON1 status (QQ genotype and lowered CMPAase activity) and lowered IgM to MDA. Partial least squares bootstrapping shows that 41.0% of the variance in negative symptoms, psychosis, hostility, excitation, mannerism, psychomotor retardation, and formal thought disorders was explained by increased TNF-α and PON1 status (QQ genotype and lowered CMPAase activity), which lowered IL-4 and IgM to MDA as well as male sex and lowered education. We found that 47.9% of the variance in verbal fluency, word list memory, true recall, Mini-Mental State Examination, and executive functions was predicted by increased TNF-α and lowered IL-4, IgM to MDA, and education. In addition, both TNF-α and IL-4 levels were significantly associated with lowered IgM to MDA, while TNF-α was correlated with PON1 status. These data provide evidence that the symptomatic (both the deficit subtype and OSOS) and cognitive impairments in schizophrenia are to a large extent mediated by the effects of immune-mediated neurotoxicity as well as lowered regulation by the innate immune system.
Sigma-Aldrich
4-(Chloromethyl)phenyl acetate, 98%
Pricing and availability is not currently available.