Merck
Megalin: A Novel Endocytic Receptor for Prorenin and Renin.
Hypertension (Dallas, Tex. : 1979) (2020-03-24)
Yuan Sun, Alexandre Goes Martini, Manoe J Janssen, Ingrid M Garrelds, Rosalinde Masereeuw, Xifeng Lu, A H Jan Danser
ABSTRACT
Megalin is an endocytic receptor contributing to protein reabsorption. Impaired expression or trafficking of megalin increases urinary renin and allowed the detection of prorenin, which normally is absent in urine. Here, we investigated (pro)renin uptake by megalin, using both conditionally immortalized proximal tubule epithelial cells and Brown Norway Rat yolk sac cells (BN16). To distinguish binding and internalization, cells were incubated with recombinant human (pro)renin at 4°C and 37°C, respectively. (Pro)renin levels were assessed by immunoradiometric assay. At 4°C, BN16 cells bound 3× more prorenin than renin, suggestive for a higher affinity of prorenin. Similarly, at 37°C, prorenin accumulated at 3- to 4-fold higher levels than renin in BN16 cells. Consequently, depletion of medium prorenin (but not renin) content occurred after 24 hours. No such differences were observed in conditionally immortalized proximal tubule epithelial cells, and M6P (mannose-6-phosphate) greatly reduced conditionally immortalized proximal tubule epithelial cells (pro)renin uptake, suggesting that these cells accumulate (pro)renin largely via M6P receptors. M6P did not affect (pro)renin uptake in BN16 cells. Yet, inhibiting megalin expression with siRNA greatly reduced (pro)renin binding and internalization by BN16 cells. Furthermore, treating BN16 cells with albumin, an endogenous ligand of megalin, also decreased binding and internalization of (pro)renin, while deleting the (pro)renin receptor affected the latter only. Exposing prorenin's prosegment with the renin inhibitor aliskiren dramatically increased prorenin binding, while after prosegment cleavage with trypsin prorenin binding was identical to that of renin. In conclusion, megalin might function as an endocytic receptor for (pro)renin and displays a preference for prorenin. Megalin-mediated endocytosis requires the (pro)renin receptor.
Sigma-Aldrich
MISSION® esiRNA, targeting human ATP6AP2