Merck
  • Increased intracellular survival of Salmonella Typhimurium ST313 in HIV-1-infected primary human macrophages is not associated with Salmonella hijacking the HIV compartment.
Increased intracellular survival of Salmonella Typhimurium ST313 in HIV-1-infected primary human macrophages is not associated with Salmonella hijacking the HIV compartment.
Biology of the cell (2020-01-11)
G Lê-Bury, C Deschamps, C Kizilyaprak, W Blanchard, J Daraspe, A Dumas, M A Gordon, J C D Hinton, B M Humbel, F Niedergang
ABSTRACT
Non-typhoidal Salmonella (NTS) causes a severe invasive syndrome (iNTS disease) described in HIV-positive adults. The impact of HIV-1 on Salmonella pathogenesis and the molecular basis for the differences between these bacteria and classical diarrhoeal S. Typhimurium remains unclear. Here, we show that iNTS-associated S. Typhimurium Sequence Type 313 (ST313) bacteria show greater intracellular survival in primary human macrophages, compared with a 'classical' diarrhoeal S. Typhimurium ST19 isolate. The increased intracellular survival phenotype of ST313 is more pronounced in HIV-infected macrophages. We explored the possibility that the bacteria take advantage of the HIV-associated viral-containing compartments created in human macrophages that have low pH. Confocal fluorescence microscopy and focussed ion beam-scanning electron microscopy tomography showed that Salmonella did not co-localise extensively with HIV-positive compartments. The capacity of ST313 bacteria to survive better than ST19 bacteria within primary human macrophages is enhanced in cells pre-infected with HIV-1. Our results indicate that the ST313 bacteria do not directly benefit from the niche created by the virus in HIV-1-infected macrophages, and that they might take advantage from a more globally modified host cell. A better understanding of the interplay between HIV-1 and Salmonella is important not only for these bacteria but also for other opportunistic pathogens.
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Pricing and availability is not currently available.
Sigma-Aldrich
Epoxy Embedding Medium kit, embedding resin for electron microscopy
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphate Buffer Solution, 1.0 M, pH 7.4 (25 °C)
Pricing and availability is not currently available.