Merck
  • Slit2 May Underlie Divergent Induction by Thyrotropin of IL-23 and IL-12 in Human Fibrocytes.
Slit2 May Underlie Divergent Induction by Thyrotropin of IL-23 and IL-12 in Human Fibrocytes.
Journal of immunology (Baltimore, Md. : 1950) (2020-02-23)
Roshini Fernando, Stephen J Atkins, Terry J Smith
ABSTRACT
IL-23 and IL-12, two structurally related heterodimeric cytokines sharing a common subunit, divergently promote Th cell development and expansion. Both cytokines have been implicated in the pathogenesis of thyroid-associated ophthalmopathy (TAO), an autoimmune component of Graves disease. In TAO, CD34+ fibrocytes, putatively derived from bone marrow, can be identified in the orbit. There they masquerade as CD34+ orbital fibroblasts (OF) (CD34+ OF) and cohabitate with CD34- OF in a mixed fibroblast population (GD-OF). Slit2, a neural axon repellent, is expressed and released by CD34- OF and dampens the inflammatory phenotype of fibrocytes and CD34+ OF. In this study we report that thyrotropin (TSH) and the pathogenic, GD-specific monoclonal autoantibody, M22, robustly induce IL-23 in human fibrocytes; however, IL-12 expression is essentially undetectable in these cells under basal conditions or following TSH-stimulation. In contrast, IL-12 is considerably more inducible in GD-OF, cells failing to express IL-23. This divergent expression and induction of cytokines appears to result from cell type-specific regulation of both gene transcription and mRNA stabilities. It appears that the JNK pathway activity divergently attenuates IL-23p19 expression while enhancing that of IL-12p35. The shift from IL-23p19 expression in fibrocytes to that of IL-12p35 in their derivative CD34+ OF results from the actions of Slit2. Thus, Slit2 might represent a molecular determinant of balance between IL-23 and IL-12 expression, potentially governing immune responses in TAO.
Sigma-Aldrich
Thymidine 5′-triphosphate sodium salt, ≥96%
Millipore
Accutase cell detachment solution, A cell detachment solution of proteolytic & collagenolytic enzymes. The reagent is useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.
Sigma-Aldrich
SLIT-2-N human, recombinant, expressed in HEK 293 cells, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside