Merck
  • FlhF, a signal recognition particle-like GTPase, is involved in the regulation of flagellar arrangement, motility behaviour and protein secretion in Bacillus cereus.
FlhF, a signal recognition particle-like GTPase, is involved in the regulation of flagellar arrangement, motility behaviour and protein secretion in Bacillus cereus.
Microbiology (Reading, England) (2007-07-31)
Sara Salvetti, Emilia Ghelardi, Francesco Celandroni, Mara Ceragioli, Francesco Giannessi, Sonia Senesi
ABSTRACT
Flagellar arrangement is a highly conserved feature within bacterial species. However, only a few genes regulating cell flagellation have been described in polar flagellate bacteria. This report demonstrates that the arrangement of flagella in the peritrichous flagellate Bacillus cereus is controlled by flhF. Disruption of flhF in B. cereus led to a reduction in the number of flagella from 10-12 to 1-3 filaments per cell in the insertion mutant MP06. Moreover, compared to the parental strain, MP06 exhibited: (i) shorter smooth swimming phases, causing reduced swimming motility but not affecting chemotaxis; (ii) complete inhibition of swarming motility, as differentiated swarm cells were never detected; (iii) an increased amount of extracellular proteins; and (iv) differential export of virulence determinants, such as haemolysin BL (HBL), phosphatidylcholine-preferring phospholipase C (PC-PLC) and non-haemolytic enterotoxin (NHE). Introduction of a plasmid harbouring flhF (pDGflhF) into MP06 completely restored the wild-type phenotype in the trans-complemented strain MP07. B. cereus flhF was found to constitute a monocistronic transcriptional unit and its overexpression did not produce abnormal features in the wild-type background. Characterization of a B. cereus mutant (MP05) carrying a partial flhF deletion indicated that the last C-terminal domain of FlhF is involved in protein export while not required for flagellar arrangement and motility behaviour. Taken together, these data suggest that B. cereus FlhF is a promising candidate for connecting diverse cellular functions, such as flagellar arrangement, motility behaviour, pattern of protein secretion and virulence phenotype.
Sigma-Aldrich
Phospholipase C from Bacillus cereus, ≥200 units/mg protein
Pricing and availability is not currently available.