Merck
  • Inhibition of fructose 1,6-bisphosphatase reduces excessive endogenous glucose production and attenuates hyperglycemia in Zucker diabetic fatty rats.
Inhibition of fructose 1,6-bisphosphatase reduces excessive endogenous glucose production and attenuates hyperglycemia in Zucker diabetic fatty rats.
Diabetes (2006-05-30)
Paul D van Poelje, Scott C Potter, Visvanathan C Chandramouli, Bernard R Landau, Qun Dang, Mark D Erion
ABSTRACT
Gluconeogenesis is increased in type 2 diabetes and contributes significantly to fasting and postprandial hyperglycemia. We recently reported the discovery of the first potent and selective inhibitors of fructose 1,6-bisphosphatase (FBPase), a rate-controlling enzyme of gluconeogenesis. Herein we describe acute and chronic effects of the lead inhibitor, MB06322 (CS-917), in rodent models of type 2 diabetes. In fasting male ZDF rats with overt diabetes, a single dose of MB06322 inhibited gluconeogenesis by 70% and overall endogenous glucose production by 46%, leading to a reduction in blood glucose of >200 mg/dl. Chronic treatment of freely feeding 6-week-old male Zucker diabetic fatty (ZDF) rats delayed the development of hyperglycemia and preserved pancreatic function. Elevation of lactate ( approximately 1.5-fold) occurred after 4 weeks of treatment, as did the apparent shunting of precursors into triglycerides. Profound glucose lowering ( approximately 44%) and similar metabolic ramifications were associated with 2-week intervention therapy of 10-week-old male ZDF rats. In high-fat diet-fed female ZDF rats, MB06322 treatment for 2 weeks fully attenuated hyperglycemia without evidence of metabolic perturbation other than a modest reduction in glycogen stores ( approximately 20%). The studies confirm that excessive gluconeogenesis plays an integral role in the pathophysiology of type 2 diabetes and suggest that FBPase inhibitors may provide a future treatment option.
Sigma-Aldrich
D-Fructose 1,6-bisphosphate tetra(cyclohexylammonium) salt, ≥95% anhydrous basis (enzymatic)
Pricing and availability is not currently available.