Merck
pH-Dependent Interaction between C-Peptide and Phospholipid Bicelles.
Journal of biophysics (Hindawi Publishing Corporation : Online) (2012-08-01)
Sofia Unnerståle, Lena Mäler
ABSTRACT
C-peptide is the connecting peptide between the A and B chains of insulin in proinsulin. In this paper, we investigate the interaction between C-peptide and phospholipid bicelles, by circular dichroism and nuclear magnetic resonance spectroscopy, and in particular the pH dependence of this interaction. The results demonstrate that C-peptide is largely unstructured independent of pH, but that a weak structural induction towards a short stretch of β-sheet is induced at low pH, corresponding to the isoelectric point of the peptide. Furthermore, it is demonstrated that C-peptide associates with neutral phospholipid bicelles as well as acidic phospholipid bicelles at this low pH. C-peptide does not undergo a large structural rearrangement as a consequence of lipid interaction, which indicates that the folding and binding are uncoupled. In vivo, local variations in environment, including pH, may cause C-peptide to associate with lipids, which may affect the aggregation state of the peptide.
Product Description
Avanti
06:0 PC-d22, Avanti Research - A Croda Brand 790427P, powder
Pricing and availability is not currently available.