Merck
  • Schizorhodopsins: A family of rhodopsins from Asgard archaea that function as light-driven inward H+ pumps.
Schizorhodopsins: A family of rhodopsins from Asgard archaea that function as light-driven inward H+ pumps.
Science advances (2020-04-18)
Keiichi Inoue, Satoshi P Tsunoda, Manish Singh, Sahoko Tomida, Shoko Hososhima, Masae Konno, Ryoko Nakamura, Hiroki Watanabe, Paul-Adrian Bulzu, Horia L Banciu, Adrian-Ştefan Andrei, Takayuki Uchihashi, Rohit Ghai, Oded Béjà, Hideki Kandori
ABSTRACT
Schizorhodopsins (SzRs), a rhodopsin family first identified in Asgard archaea, the archaeal group closest to eukaryotes, are present at a phylogenetically intermediate position between typical microbial rhodopsins and heliorhodopsins. However, the biological function and molecular properties of SzRs have not been reported. Here, SzRs from Asgardarchaeota and from a yet unknown microorganism are expressed in Escherichia coli and mammalian cells, and ion transport assays and patch clamp analyses are used to demonstrate SzR as a novel type of light-driven inward H+ pump. The mutation of a cytoplasmic glutamate inhibited inward H+ transport, suggesting that it functions as a cytoplasmic H+ acceptor. The function, trimeric structure, and H+ transport mechanism of SzR are similar to that of xenorhodopsin (XeR), a light-driven inward H+ pumping microbial rhodopsins, implying that they evolved convergently. The inward H+ pump function of SzR provides new insight into the photobiological life cycle of the Asgardarchaeota.
Sigma-Aldrich
Anti-c-Myc antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Membrane Scaffold Protein 1E3D1, recombinant, expressed in E. coli
Avanti
16:0-18:1 PG, Avanti Research - A Croda Brand
Sigma-Aldrich
Soy Phospholipids, mixture of phospholipids
Avanti
16:0-18:1 PE, Avanti Research - A Croda Brand