- Topical ROR Inverse Agonists Suppress Inflammation in Mouse Models of Atopic Dermatitis and Acute Irritant Dermatitis.
Topical ROR Inverse Agonists Suppress Inflammation in Mouse Models of Atopic Dermatitis and Acute Irritant Dermatitis.
The retinoic acid receptor-related orphan receptors RORα and RORγ are critical for the functions of specific subsets of T cells and innate lymphoid cells, which are key drivers of inflammatory disease in barrier tissues. Here, we investigate the anti-inflammatory potential of SR1001, a synthetic RORα/γ inverse agonist, in mouse models of atopic dermatitis and acute irritant dermatitis. Topical treatment with SR1001 reduces epidermal and dermal features of MC903-induced atopic dermatitis-like disease and suppresses the production of type 2 cytokines and other inflammatory mediators in lesional skin. In the epidermis, SR1001 treatment blocks MC903-induced expression of TSLP and reverses impaired keratinocyte differentiation. SR1001 is also effective in alleviating acute dermatitis triggered by 12-O-tetradecanoylphorbol-13-acetate. Overall, our results suggest that RORα/γ are important therapeutic targets for cutaneous inflammation and suggest topical usage of inhibitory ligands as an approach to treating skin diseases of inflammatory etiology.