Merck
  • Anisotropic active ligandations in siRNA-Loaded hybrid nanodiscs lead to distinct carcinostatic outcomes by regulating nano-bio interactions.
Anisotropic active ligandations in siRNA-Loaded hybrid nanodiscs lead to distinct carcinostatic outcomes by regulating nano-bio interactions.
Biomaterials (2020-05-11)
Qing Chen, Guannan Guan, Feiyang Deng, Dan Yang, Peiyao Wu, Shuangming Kang, Ruimeng Sun, Xiaoyou Wang, Demin Zhou, Wenbing Dai, Xueqing Wang, Hua Zhang, Bing He, Dawei Chen, Qiang Zhang
ABSTRACT
Active targeting modification is one of the foremost nanomedicine strategies for the efficacy improvement. Compared to the homogeneous ligandation on spherical nanocarriers, non-spherical nanomedicines usually make the ligand modification more complicated. The modified ligands always exhibit anisotropy and heterogeneity. However, there is very little systematic study on these diversified anisotropic modifications. The efficacy difference and underlying mechanism were still unclear. Here, we separately fabricated hybrid nanodiscs (NDs) conjugated with cRGD on the edge and plane surfaces to engineer two anisotropic targeting nanocarriers (E-cRGD-NDs and P-cRGD-NDs, respectively) for gene delivery. The ligand anisotropy endowed NDs with diversified cellular interactions, and caused different efficacies between E-cRGD-NDs and P-cRGD-NDs. Of note, E-cRGD-NDs showed significant superiority in siRNA loading, cellular uptake, silence efficiency, protein expression and even in vivo efficacy. The mechanism investigation revealed the functional anisotropy specifically for E-cRGD-NDs. The edge modification of cRGD efficiently separated the targeting and siRNA loading domains, maximizing their respective functions. These findings reflected the unique effect of ligand anisotropy, also provided a new strategy for the targeting screening of extensive nanomedicines.
Product Description
Avanti
07:0 PC (DHPC), Avanti Research - A Croda Brand
Avanti
18:1 TAP (DOTAP), Avanti Research - A Croda Brand