Merck
  • Chlamydia pneumoniae infection promotes vascular smooth muscle cell migration via c-Fos/interleukin-17C signaling.
Chlamydia pneumoniae infection promotes vascular smooth muscle cell migration via c-Fos/interleukin-17C signaling.
International journal of medical microbiology : IJMM (2019-09-09)
Ningbo Zheng, Lijun Zhang, Beibei Wang, Guangyan Wang, Jingya Liu, Guolin Miao, Xi Zhao, Changle Liu, Lijun Zhang
ABSTRACT
Chlamydia pneumoniae (C. pneumoniae) infection is associated with the initiation and progression of atherosclerosis. The migration of vascular smooth muscle cell (VSMC) from the media to the intima is a key event in the development of atherosclerosis. Interleukin-17C (IL-17C) could enhance cell migration ability. The aim of our study is to investigate the role of IL-17C in C. pneumoniae infection-promoted VSMC migration, thereby possibly accelerating atherosclerosis. We firstly demonstrated that C. pneumoniae infection significantly increased IL-17C expression in VSMCs in the atherosclerotic lesion area from ApoE deficient mice. Our in vitro study further showed that IL-17C is required for C. pneumoniae infection-promoted VSMC migration, and its expression could be regulated by c-Fos through phosphorylating extracellular signal-regulated kinase (ERK). Unexpectedly, in the present study, we also found that IL-17C is critical for C. pneumoniae infection-induced c-Fos activation. c-Fos expression and activation induced by the exposure to recombinant IL-17C were markedly suppressed in the presence of the ERK inhibitor PD98059. These results suggest a possible positive feedback between c-Fos and IL-17C after C. pneumoniae infection. Taken together, our results indicate that C. pneumoniae infection promotes VSMC migration via c-Fos/IL-17C signaling.
Sigma-Aldrich
MISSION® esiRNA, targeting human FOS