- The neuropeptide calcitonin gene-related peptide (CGRP) stimulates T cell migration into collagen matrices.
The neuropeptide calcitonin gene-related peptide (CGRP) stimulates T cell migration into collagen matrices.
CGRP significantly stimulated migration of non-activated and anti-CD3 activated T lymphocytes into a collagen matrix when present inside the collagen, whereas somatostatin-14, NPY, substance P, VIP, beta-endorphin and metenkephalin had no or little effect. The CGRP antagonist CGRP 8-37 abrogated the CGRP-induced cell infiltration. Virtually all migrating cells were CD3+ (>96%) and CGRP did not stimulate B-cell migration. The migration capacity showed no selective relationship to the expression of CD4+, CD8+, CD45RO+ (memory), or CD45RA+ (naive) T cell markers indicating that the regulation of T cell migration is distinct from that of the major T cell phenotypes.