Merck
  • Herbivore-specific plant volatiles prime neighboring plants for nonspecific defense responses.
Herbivore-specific plant volatiles prime neighboring plants for nonspecific defense responses.
Plant, cell & environment (2019-11-24)
Bipana Paudel Timilsena, Irmgard Seidl-Adams, James H Tumlinson
ABSTRACT
Plants produce species-specific herbivore-induced plant volatiles (HIPVs) after damage. We tested the hypothesis that herbivore-specific HIPVs prime neighboring plants to induce defenses specific to the priming herbivore. Since Manduca sexta (specialist) and Heliothis virescens (generalist) herbivory induced unique HIPV profiles in Nicotiana benthamiana, we used these HIPVs to prime receiver plants for defense responses to simulated herbivory (mechanical wounding and herbivore regurgitant application). Jasmonic acid (JA) accumulations and emitted volatile profiles were monitored as representative defense responses since JA is the major plant hormone involved in wound and defense signaling and HIPVs have been implicated as signals in tritrophic interactions. Herbivore species-specific HIPVs primed neighboring plants, which produced 2 to 4 times more volatiles and JA after simulated herbivory when compared to similarly treated constitutive volatile-exposed plants. However, HIPV-exposed plants accumulated similar amounts of volatiles and JA independent of the combination of priming or challenging herbivore. Furthermore, volatile profiles emitted by primed plants depended only on the challenging herbivore species but not on the species-specific HIPV profile of damaged emitter plants. This suggests that feeding by either herbivore species primed neighboring plants for increased HIPV emissions specific to the subsequently attacking herbivore and is probably controlled by JA.
Sigma-Aldrich
Nonyl acetate, ≥97%, FCC
Pricing and availability is not currently available.
Sigma-Aldrich
trans-2-Hexen-1-al, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Myrcene, sum of isomers, ≥90%, natural, FG
Pricing and availability is not currently available.