Merck
  • Silicone Oil Particles in Prefilled Syringes With Human Monoclonal Antibody, Representative of Real-World Drug Products, Did Not Increase Immunogenicity in In Vivo and In Vitro Model Systems.
Silicone Oil Particles in Prefilled Syringes With Human Monoclonal Antibody, Representative of Real-World Drug Products, Did Not Increase Immunogenicity in In Vivo and In Vitro Model Systems.
Journal of pharmaceutical sciences (2019-10-20)
Nathan H Joh, Lisa Thomas, Twinkle R Christian, Alla Verlinsky, Nancy Jiao, Nilo Allotta, Vibha Jawa, Shawn Cao, Linda O Narhi, Marisa K Joubert
ABSTRACT
Silicone oil is a lubricant for prefilled syringes (PFS), a common primary container for biotherapeutics. Silicone oil particles (SiOP) shed from PFS are a concern for patients due to their potential for increased immunogenicity and therefore also of regulatory concern. To address the safety concern in a context of manufacturing and distribution of drug product (DP), SiOP was increased (up to ∼25,000 particles/mL) in PFS filled with mAb1, a fully human antibody drug, by simulated handling of DP mimicked by drop shock. These samples are characterized in a companion report (Jiao N et al. J Pharm Sci. 2020). The risk of immunogenicity was then assessed using in vitro and in vivo immune model systems. The impact of a common DP excipient, polysorbate 80, on both the formation and biological consequences of SiOP was also tested. SiOP was found associated with (1) minimal cytokine secretion from human peripheral blood mononuclear cells, (2) no response in cell lines that report NF-κB/AP-1 signaling, and (3) no antidrug antibodies or significant cytokine production in transgenic Xeno-het mice, whether or not mAb1 or polysorbate 80 was present. These results suggest that SiOP in mAb1, representative of real-world DP in PFS, poses no increased risk of immunogenicity.
Sigma-Aldrich
Anti-Mouse IgG (Fc specific) antibody produced in goat, 2.0 mg/mL, affinity isolated antibody