Merck
  • An unexpected antioxidant and redox activity for the classic copper-chelating drug penicillamine.
An unexpected antioxidant and redox activity for the classic copper-chelating drug penicillamine.
Free radical biology & medicine (2019-12-21)
Li Mao, Chun-Hua Huang, Jie Shao, Li Qin, Dan Xu, Bo Shao, Ben-Zhan Zhu
ABSTRACT
Penicillamine has been widely-used clinically as a copper-chelating drug for the treatment of copper-overload in Wilson's disease. In this study, we found that penicillamine provided marked protection against cytotoxicity induced by tetrachlorohydroquinone (TCHQ), a major toxic metabolite of the well-known wood preservative pentachlorophenol, while other classic copper-chelating agents do not. We found, unexpectedly, that both TCHQ autooxidation and tetrachlorosemiquinone radical (TCSQ•-) formation were remarkably delayed by penicillamine. Further investigation showed that TCSQ•- was reduced back to TCHQ by penicillamine, with the concurrent formation of its corresponding disulfide. These data demonstrated that the protection by penicillamine against TCHQ-induced toxicity was not due to its classic Cu-chelating property, but rather to its reduction of the reactive TCSQ•- to the much less-reactive TCHQ. This is the first report of an unexpected antioxidant and redox activity for penicillamine, which might prove highly relevant to its biological activities.
Sigma-Aldrich
Tetrafluorohydroquinone, 95%
Sigma-Aldrich
D-Penicillamine, 98-101%
Sigma-Aldrich
D-Penicillamine disulfide, 97%
Sigma-Aldrich
DL-Cysteine, technical grade