Merck
  • Highly multiplexed single-cell RNA-seq by DNA oligonucleotide tagging of cellular proteins.
Highly multiplexed single-cell RNA-seq by DNA oligonucleotide tagging of cellular proteins.
Nature biotechnology (2019-12-25)
Jase Gehring, Jong Hwee Park, Sisi Chen, Matthew Thomson, Lior Pachter
ABSTRACT
We describe a universal sample multiplexing method for single-cell RNA sequencing in which fixed cells are chemically labeled by attaching identifying DNA oligonucleotides to cellular proteins. Analysis of a 96-plex perturbation experiment revealed changes in cell population structure and transcriptional states that cannot be discerned from bulk measurements, establishing an efficient method for surveying cell populations from large experiments or clinical samples with the depth and resolution of single-cell RNA sequencing.
Sigma-Aldrich
Neural Stem Cell Basal Medium, The Neural Stem Cell Basal Medium is a defined serum-free, growth factor-free medium that has been optimized for the growth & in vitro differentiation of neural stem cells derived from rodents.